siRNA Details
Gene Symbol: Upk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108911.1 | NP_001102381.1 | 1 | 183 | ||
XM_006228782.2 | XP_006228844.1 | 3 | 540 | |||
XM_006228784.2 | XP_006228846.1 | 3 | 540 | |||
XM_008759176.1 | XP_008757398.1 | 2 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
