siRNA Details
Gene Symbol: Rrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013236.1 | NP_001013254.1 | 15, 16 | 1974 | ||
GenBank mRNA | BC085906.1 | - | 15, 16 | 1974 | 7193834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
