siRNA Details
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014244.1 | NP_001014266.1 | 5 | 470 | ||
XM_008759774.1 | XP_008757996.1 | 3 | 1608 | |||
XM_008759775.1 | XP_008757997.1 | 4 | 1511 | |||
XM_008759776.1 | XP_008757998.1 | 3 | 1608 | |||
XM_008759777.1 | XP_008757999.1 | 3 | 1608 | |||
XM_008759778.1 | XP_008758000.1 | 4 | 926 | |||
XM_008759779.1 | XP_008758001.1 | 4 | 1083 | |||
XM_008759780.1 | XP_008758002.1 | 2 | 112 | |||
XM_008759781.1 | XP_008758003.1 | 5 | 407 | |||
XM_008759782.1 | XP_008758004.1 | 3 | 1608 | |||
GenBank mRNA | BC079235.1 | - | 5 | 470 | 7132617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
