siRNA Details
Gene Symbol: Tmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012216.1 | NP_001012216.1 | 7 | 1304 | ||
GenBank mRNA | BC088406.1 | - | 7 | 1304 | 7112487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
