siRNA Details
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276487.1 | NP_001263416.1 | 3 | 276 | ||
XM_006230132.2 | XP_006230194.1 | 2 | 216 | |||
XM_006230133.1 | XP_006230195.1 | 2 | 212 | |||
XM_006230134.2 | XP_006230196.2 | 3 | 2519 | |||
GenBank mRNA | BC166909.1 | - | 3 | 273 | 6889767 |
