siRNA Details
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276487.1 | NP_001263416.1 | 2 | 211 | ||
XM_006230132.2 | XP_006230194.1 | 1 | 151 | |||
XM_006230133.1 | XP_006230195.1 | 1 | 147 | |||
XM_006230134.2 | XP_006230196.2 | 2 | 2454 | |||
XM_008759783.1 | XP_008758005.1 | 2 | 2454 | |||
GenBank mRNA | BC166909.1 | - | 2 | 208 | 6889767 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001285960, NM_029610, NM_001285959, NM_001285961, AK009135, and others.
