siRNA Details
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014245.2 | NP_001014267.1 | 3 | 372 | ||
XM_006231061.2 | XP_006231123.1 | 5 | 527 | |||
XM_006231062.2 | XP_006231124.1 | 4 | 515 | |||
XM_008760238.1 | XP_008758460.1 | 4 | 446 | |||
XM_008760239.1 | XP_008758461.1 | 4 | 446 | |||
XM_008760240.1 | XP_008758462.1 | 3 | 333 | |||
GenBank mRNA | BC083864.1 | - | 3 | 375 | 7127736 |

More Information
Other Species Targeted

Gene Ontology Categories:
