siRNA Details
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014245.2 | NP_001014267.1 | 1, 2 | 119 | ||
XM_006231061.2 | XP_006231123.1 | 2, 3 | 200 | |||
XM_006231062.2 | XP_006231124.1 | 2, 3 | 262 | |||
XM_008760238.1 | XP_008758460.1 | 2, 3 | 193 | |||
XM_008760239.1 | XP_008758461.1 | 2, 3 | 193 | |||
GenBank mRNA | BC083864.1 | - | 1, 2 | 122 | 7127736 |
