siRNA Details
Gene Symbol: Cfap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231588.2 | XP_006231650.1 | 30 | 3711 | ||
XM_008760497.1 | XP_008758719.1 | 33 | 4349 | |||
XM_345041.7 | XP_345042.5 | 34 | 4499 | |||
GenBank mRNA | BC092207.1 | - | 9 | 1008 |
