siRNA Details
Gene Symbol: Cacul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014248.1 | NP_001014270.1 | 2 | 732 | ||
XM_006231678.2 | XP_006231740.1 | 2 | 892 | |||
GenBank mRNA | BC083839.1 | - | 2 | 732 | 7114271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
