siRNA Details
Gene Symbol: Micu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199412.1 | NP_955444.1 | 10 | 1187 | ||
XM_006256420.2 | XP_006256482.1 | 10 | 1302 | |||
XM_006256421.2 | XP_006256483.1 | 11 | 1295 | |||
XM_006256422.2 | XP_006256484.1 | 11 | 1289 | |||
GenBank mRNA | BC062075.1 | - | 10 | 1187 | 5599430 |
