siRNA Details
Gene Symbol: Dse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108933.1 | NP_001102403.1 | 3 | 977 | ||
XM_006256382.2 | XP_006256444.1 | 3 | 732 | |||
XM_008772900.1 | XP_008771122.1 | 3 | 809 | |||
XM_008772901.1 | XP_008771123.1 | 5 | 1311 | |||
GenBank mRNA | BC168891.1 | - | 5 | 1245 | 7110121 |
