siRNA Details
Gene Symbol: Pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014249.1 | NP_001014271.1 | 3 | 724 | ||
XM_008773004.1 | XP_008771226.1 | 3 | 796 | |||
GenBank mRNA | BC085898.1 | - | 3 | 724 | 7193369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
