siRNA Details
Gene Symbol: Bves | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077590.1 | NP_001071058.1 | 4 | 582 | ||
GenBank mRNA | AY786529.2 | - | 4 | 582 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024285, AF204174, BC132044, BC132018, AF124510.
Gene Ontology Categories:
