siRNA Details
Gene Symbol: Col4a3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108935.1 | NP_001102405.1 | 17 | 2240 | ||
XM_006231827.2 | XP_006231889.1 | 16 | 2158 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Col4a3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108935.1 | NP_001102405.1 | 17 | 2240 | ||
XM_006231827.2 | XP_006231889.1 | 16 | 2158 |