siRNA Details
Gene Symbol: Marveld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108936.1 | NP_001102406.1 | 7 | 1781 | ||
XM_008760679.1 | XP_008758901.1 | 7 | 1716 | |||
XM_008760680.1 | XP_008758902.1 | 7 | 1850 | |||
XM_008760681.1 | XP_008758903.1 | 7 | 1818 |
