siRNA Details
Gene Symbol: Trim55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012218.1 | NP_001012218.1 | 5 | 860 | ||
XM_006232181.2 | XP_006232243.1 | 5 | 898 | |||
XM_006232182.2 | XP_006232244.1 | 5 | 898 | |||
GenBank mRNA | BC087100.1 | - | 5 | 860 | 7192411 |
