siRNA Details
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108941.1 | NP_001102411.1 | 9 | 1045 | ||
XM_006232190.2 | XP_006232252.1 | 8 | 938 | |||
XM_008760889.1 | XP_008759111.1 | 9 | 1801 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030732, AK136255, AK036064, AK033347, AK029595.
Gene Ontology Categories:
