siRNA Details
Gene Symbol: Bbs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232287.2 | XP_006232349.1 | 1 | 1911 | ||
XM_006232288.2 | XP_006232350.1 | 1 | 1911 | |||
XM_006232289.2 | XP_006232351.1 | 1 | 1911 |
