siRNA Details
Gene Symbol: Ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126080.2 | NP_001119552.1 | 4, 5 | 242 | ||
GenBank mRNA | BC091395.1 | - | 4, 5 | 178 | 7323172 | |
CB787557.1 | - | 4, 5 | 280 | |||
CV112030.1 | - | 3, 4 | 132 | |||
FQ229705.1 | - | 4, 5 | 213 |

More Information
Other Species Targeted

Gene Ontology Categories:
