siRNA Details
Gene Symbol: Tuft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108948.1 | NP_001102418.1 | 9 | 941 | ||
XM_006232890.2 | XP_006232952.1 | 13 | 1320 | |||
XM_006232891.2 | XP_006232953.1 | 12 | 1245 | |||
GenBank mRNA | BC168687.1 | - | 1178 | 7932385 |
