siRNA Details
Gene Symbol: Pip5k1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042621.1 | NP_001036086.1 | 2, 3 | 434 | ||
XM_008761315.1 | XP_008759537.1 | 2, 3 | 563 | |||
XM_008761316.1 | XP_008759538.1 | 2, 3 | 560 | |||
GenBank mRNA | AY724476.1 | - | 2, 3 | 434 |
