siRNA Details
Gene Symbol: Hormad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108949.1 | NP_001102419.1 | 9 | 551 | ||
XM_006232951.2 | XP_006233013.1 | 8 | 506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
