siRNA Details
Gene Symbol: Hormad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108949.1 | NP_001102419.1 | 9 | 536 | ||
XM_006232951.2 | XP_006233013.1 | 8 | 491 |

More Information
Other Species Targeted

Gene Ontology Categories:
