siRNA Details
Gene Symbol: Ankrd35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232996.2 | XP_006233058.1 | 9 | 2599 | ||
XM_345258.7 | XP_345259.5 | 11 | 2869 | |||
GenBank mRNA | FQ221348.1 | - | 10 | 2773 |
