siRNA Details
Gene Symbol: Hipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100986.1 | NP_001094456.1 | 6 | 1738 | ||
XM_006233090.2 | XP_006233152.1 | 6 | 1689 | |||
XM_006233097.2 | XP_006233159.1 | 6 | 1688 | |||
XM_008761412.1 | XP_008759634.1 | 6 | 1699 | |||
XM_008761413.1 | XP_008759635.1 | 6 | 2398 | |||
XM_008761414.1 | XP_008759636.1 | 7 | 2553 | |||
XM_008761415.1 | XP_008759637.1 | 7 | 2884 | |||
XM_008761416.1 | XP_008759638.1 | 7 | 2223 | |||
XM_008761417.1 | XP_008759639.1 | 6 | 1901 | |||
XM_008761418.1 | XP_008759640.1 | 6 | 2003 | |||
XM_008761419.1 | XP_008759641.1 | 6 | 1700 | |||
XM_008761420.1 | XP_008759642.1 | 6 | 2393 | |||
GenBank mRNA | EF460311.1 | - | 6 | 1738 | ||
EF460312.1 | - | 6 | 1738 |
