siRNA Details
Gene Symbol: Alx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007012.1 | NP_001007013.1 | 2 | 530 | ||
GenBank mRNA | AY488087.1 | - | 2 | 530 |

More Information
Other Species Targeted

Gene Ontology Categories:
