siRNA Details
Gene Symbol: Gnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108950.2 | NP_001102420.2 | 4 | 282 | ||
XM_006233171.1 | XP_006233233.1 | 4 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
