siRNA Details
Gene Symbol: Tmem56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135879.1 | NP_001129351.1 | 4 | 371 | ||
XM_006233244.2 | XP_006233306.1 | 6 | 645 | |||
XM_006233246.2 | XP_006233308.1 | 6 | 648 | |||
XM_006233251.2 | XP_006233313.1 | 6 | 468 | |||
XM_006233252.2 | XP_006233314.1 | 5 | 416 | |||
XM_006233253.2 | XP_006233315.1 | 5 | 550 | |||
XM_006233254.2 | XP_006233316.1 | 5 | 553 | |||
XM_008761467.1 | XP_008759689.1 | 5 | 815 | |||
XM_008761468.1 | XP_008759690.1 | 5 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
