siRNA Details
Gene Symbol: Lcn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128183.1 | NP_001121655.1 | 2, 3 | 255 | ||
XM_006233632.2 | XP_006233694.2 | 2, 3 | 396 | |||
GenBank mRNA | DQ537491.1 | - | 2, 3 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
