siRNA Details
Gene Symbol: Trub2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014257.2 | NP_001014279.1 | 5 | 461 | ||
GenBank mRNA | BC084712.1 | - | 5 | 436 | 7303031 | |
CB613146.1 | - | 5 | 461 | |||
FQ225796.1 | - | 5 | 305 | |||
FQ231048.1 | - | 5 | 432 | |||
FQ231592.1 | - | 5 | 475 |

More Information
Other Species Targeted

Gene Ontology Categories:
