siRNA Details
Gene Symbol: Trub2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014257.2 | NP_001014279.1 | 5 | 480 | ||
GenBank mRNA | BC084712.1 | - | 5 | 455 | 7303031 | |
CB613146.1 | - | 5 | 480 | |||
FQ225796.1 | - | 5 | 324 | |||
FQ231048.1 | - | 5 | 451 | |||
FQ231592.1 | - | 5 | 494 |
