siRNA Details
Gene Symbol: Coq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031662.1 | NP_001026832.1 | 5 | 446 | ||
GenBank mRNA | BC099123.1 | - | 5 | 446 | 7462864 |

More Information
Other Species Targeted

Gene Ontology Categories:
