siRNA Details
Gene Symbol: Zbtb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108953.1 | NP_001102423.1 | 2 | 769 | ||
XM_006234101.2 | XP_006234163.1 | 2 | 865 | |||
XM_006234102.1 | XP_006234164.1 | 2 | 1140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
