siRNA Details
Gene Symbol: Olr522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000562.1 | NP_001000562.1 | 1 | 514 |
Gene Symbol: Olr528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000319.1 | NP_001000319.1 | 1 | 514 |
