siRNA Details
Gene Symbol: Nutm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108957.1 | NP_001102427.1 | 3 | 919 | ||
XM_006234708.2 | XP_006234770.1 | 3 | 915 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
