siRNA Details
Gene Symbol: Esf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100771.1 | NP_001094241.1 | 11 | 2091 | ||
XM_006235105.2 | XP_006235167.1 | 11 | 2224 | |||
XM_006235106.2 | XP_006235168.1 | 11 | 2090 | |||
GenBank mRNA | AB038233.1 | - | 11 | 2089 |
