siRNA Details
Gene Symbol: Polr3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014259.1 | NP_001014281.1 | 3 | 939 | ||
GenBank mRNA | BC089990.1 | - | 3 | 939 | 7319381 | |
FQ212572.1 | - | 3 | 961 | |||
FQ213710.1 | - | 3 | 962 | |||
FQ213830.1 | - | 3 | 973 | |||
FQ216547.1 | - | 3 | 962 | |||
FQ226947.1 | - | 3 | 944 | |||
FQ232144.1 | - | 3 | 973 | |||
U53183.1 | - | 1 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
