siRNA Details
Gene Symbol: Faxc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271065.1 | NP_001257994.1 | 1 | 367 | ||
XM_006237944.2 | XP_006238006.1 | 1 | 1115 |

More Information
Other Species Targeted

