siRNA Details
Gene Symbol: Mob3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108970.1 | NP_001102440.1 | 3 | 969 | ||
XM_006238001.2 | XP_006238063.1 | 3 | 982 | |||
XM_008763630.1 | XP_008761852.1 | 3 | 1660 | |||
XM_008763631.1 | XP_008761853.1 | 5 | 1594 | |||
XM_008763632.1 | XP_008761854.1 | 4 | 1488 | |||
XM_008763633.1 | XP_008761855.1 | 3 | 1241 | |||
XM_008763634.1 | XP_008761856.1 | 3 | 990 | |||
XM_008763635.1 | XP_008761857.1 | 5 | 2013 | |||
XM_008763636.1 | XP_008761858.1 | 3 | 1658 | |||
GenBank mRNA | FQ220583.1 | - | 2 | 460 | ||
FQ222144.1 | - | 3 | 927 |
Gene Symbol: LOC102550026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763645.1 | XP_008761867.1 | 2 | 1621 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
