siRNA Details
Gene Symbol: Igfbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108972.2 | NP_001102442.1 | 4 | 944 | ||
GenBank mRNA | BC158647.1 | - | 4 | 944 | 8364465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
