siRNA Details
Gene Symbol: Olr857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000583.1 | NP_001000583.1 | 1 | 73 |
Gene Symbol: LOC100912451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764124.1 | XP_008762346.1 | 73 |
