siRNA Details
Gene Symbol: Lsm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108976.1 | NP_001102446.1 | 2 | 329 | ||
XM_006238895.2 | XP_006238957.1 | 2 | 354 | |||
XM_006238896.2 | XP_006238958.1 | 2 | 1162 | |||
XM_006238897.2 | XP_006238959.1 | 2 | 347 | |||
GenBank mRNA | BC166902.1 | - | 2 | 289 | 6922293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
