siRNA Details
Gene Symbol: Pigv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010966.1 | NP_001010966.1 | 3 | 1284 | ||
XM_006239051.2 | XP_006239113.1 | 1 | 2049 | |||
XM_006239055.2 | XP_006239117.1 | 3 | 1030 | |||
XM_008764159.1 | XP_008762381.1 | 3 | 2463 | |||
XM_008764160.1 | XP_008762382.1 | 3 | 1007 | |||
GenBank mRNA | AB196341.1 | - | 3 | 1284 |
