siRNA Details
Gene Symbol: Epha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108977.1 | NP_001102447.1 | 5 | 1328 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Epha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108977.1 | NP_001102447.1 | 5 | 1328 |