siRNA Details
Gene Symbol: Tnfrsf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015034.1 | NP_001015034.1 | 5, 6 | 637 | ||
XM_006239586.2 | XP_006239648.1 | 6, 7 | 1522 | |||
XM_006239587.2 | XP_006239649.1 | 6, 7 | 1523 | |||
GenBank mRNA | BC091202.1 | - | 5, 6 | 637 | 7380378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
