siRNA Details
Gene Symbol: Dnajc5g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013242.1 | NP_001013260.1 | 3 | 329 | ||
XM_006239826.2 | XP_006239888.1 | 3 | 390 | |||
XM_006239827.2 | XP_006239889.1 | 3 | 390 | |||
GenBank mRNA | BC083671.1 | - | 3 | 329 | 7135668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
