siRNA Details
Gene Symbol: Cfl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108982.2 | NP_001102452.2 | 2 | 265 | ||
XM_006240104.2 | XP_006240166.1 | 2 | 352 | |||
GenBank mRNA | FQ228820.1 | - | 2 | 268 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007688, AK147174, BC007138, L29468.
Gene Ontology Categories:
