siRNA Details
Gene Symbol: Olr960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000589.1 | NP_001000589.1 | 668 |
Gene Symbol: Olr959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000486.1 | NP_001000486.1 | 1 | 668 |
