siRNA Details
Gene Symbol: Olr987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000590.1 | NP_001000590.1 | 1 | 316 |
Gene Symbol: Olr898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000058.1 | NP_001000058.1 | 1 | 316 |

More Information
Other Species Targeted

Gene Ontology Categories:
